2,6-Dimethyl-3-heptene ≥98.0% (by GC)

D1262-0.1ML
TCD1262-0.1MLEA 90.3 USD
TCD1262-0.1ML
2,6-Dimethyl-3-heptene ≥98.0% (by GC)
2,6-Dimethyl-3-heptene

CAS Number: 2738-18-3
MDL Number: MFCD00026523
Molecular Formula: C9H18
Molecular Weight: 126.24
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 15
Specific Gravity (20/20): 0.72

Formula: C₉H₁₈
MW: 126.24 g/mol
Boiling Pt: 134 °C
Density: 0.72 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00026523
CAS Number: 2738-18-3
https://us.vwr.com/stibo/web/std.lang.all/81/55/28978155.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR