N,N-Dimethylallylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
1-Dimethylamino-2-propene
, N-Allyldimethylamine
D1160-25ML
TCD1160-25MLEA
168.99
USD
TCD1160-25ML
N,N-Dimethylallylamine ≥98.0% (by GC, titration analysis)
N,N-Dimethylallylamine
CAS Number: 2155-94-4
MDL Number: MFCD00038234
Molecular Formula: C5H11N
Molecular Weight: 85.15
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): -20
Specific Gravity (20/20): 0.72
Other Product Names / Synonyms: N-Allyldimethylamine, 1-Dimethylamino-2-propene
Formula:
C₅H₁₁N MW: 85.15 g/mol Boiling Pt: 64 °C Density: 0.72 (20 °C) Flash Pt: –20 °C |
MDL Number:
MFCD00038234 CAS Number: 2155-94-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...