Chloro(dimethyl)(phenyl)silane ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
Chlorodimethylphenylsilane
CAS Number: 768-33-2
MDL Number: MFCD00000499
Molecular Formula: C8H11ClSi
Molecular Weight: 170.71
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 82
Specific Gravity (20/20): 1.03
Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis), Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Phenyldimethylchlorosilane
Formula:
C₈H₁₁ClSi MW: 170.71 g/mol Boiling Pt: 198 °C Density: 1.03 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00000499 CAS Number: 768-33-2 UN: 2987 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...