Copper(II) dimethyldithiocarbamate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dimethyldithiocarbamic Acid Copper(II) Salt

D1022-25G
TCD1022-25GEA 146.11 USD
TCD1022-25G
Copper(II) dimethyldithiocarbamate ≥98.0% (by titrimetric analysis)
Copper(II) dimethyldithiocarbamate

CAS Number: 137-29-1
MDL Number: MFCD00050845
Molecular Formula: C3H7NS2
Molecular Weight: 303.96
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Dimethyldithiocarbamic Acid Copper(II) Salt

Formula: C₃H₇NS₂
MW: 303.96 g/mol
MDL Number: MFCD00050845
CAS Number: 137-29-1
https://us.vwr.com/stibo/web/std.lang.all/62/25/28976225.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR