TCIAD1020

D1020-10G
TCD1020-10GEA 101.41 USD
TCD1020-10G
TCIAD1020
CAS Number: 829-85-6
MDL Number: MFCD00003040
Molecular Formula: C12H11P
Molecular Weight: 186.19
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 110
Specific Gravity (20/20): 1.08
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR