1,4-Diisopropylbenzene ≥98.0% (by GC)

Supplier: TCI America

D0928-25ML
TCD0928-25MLEA 52.78 USD
TCD0928-25ML
1,4-Diisopropylbenzene ≥98.0% (by GC)
1,4-Diisopropylbenzene

CAS Number: 100-18-5
MDL Number: MFCD00008892
Molecular Formula: C12H18
Molecular Weight: 162.28
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 210
Flash Point (°C): 75
Specific Gravity (20/20): 0.86

Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 210 °C
Density: 0.86 (20 °C)
Flash Pt: 75 °C
MDL Number: MFCD00008892
CAS Number: 100-18-5
https://us.vwr.com/stibo/web/std.lang.all/60/64/28976064.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR