1,1-Diphenylhydrazine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N,N-Diphenylhydrazinium chloride

D0895-25G
TCD0895-25GEA 136.98 USD
TCD0895-25G
1,1-Diphenylhydrazine hydrochloride ≥98.0% (by HPLC, titration analysis)
1,1-Diphenylhydrazine hydrochloride

CAS Number: 530-47-2
MDL Number: MFCD00012950
Molecular Formula: C12H12N2
Molecular Weight: 220.70
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 166

Formula: C₁₂H₁₃ClN₂
Melting Pt: 166 °C
MDL Number: MFCD00012950
CAS Number: 530-47-2
https://us.vwr.com/stibo/web/std.lang.all/60/15/28976015.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR