trans-2,5-Dimethylpiperazine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
D0763-25G
D0763-5G
TCD0763-25GEA
91.35
USD
TCD0763-25G
TCD0763-5G
trans-2,5-Dimethylpiperazine ≥99.0% (by GC, titration analysis)
trans-2,5-Dimethylpiperazine
CAS Number: 2815-34-1
MDL Number: MFCD00070516
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 116
Flash Point (°C): 38
Formula:
C₆H₁₄N₂ Boiling Pt: 162 °C Melting Pt: 116 °C Flash Pt: 38 °C |
MDL Number:
MFCD00070516 CAS Number: 2815-34-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...