N,N'-Dimethylhydrazine dihydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1,2-Dimethylhydrazine dihydrochloride
, sym-Dimethylhydrazine dihydrochloride
D0741-25G
TCD0741-25GEA
270.89
USD
TCD0741-25G
N,N'-Dimethylhydrazine dihydrochloride ≥98.0% (by titrimetric analysis)
N,N'-Dimethylhydrazine dihydrochloride
CAS Number: 306-37-6
MDL Number: MFCD00012522
Molecular Formula: C2H8N2
Molecular Weight: 133.02
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 168
Formula:
C₂H₁₀Cl₂N₂ Melting Pt: 168 °C |
MDL Number:
MFCD00012522 CAS Number: 306-37-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...