2,5-Dimethylfuran ≥98.0% (by GC)

Supplier: TCI America

D0725-25ML
TCD0725-25MLEA 45.87 USD
TCD0725-25ML
2,5-Dimethylfuran ≥98.0% (by GC)
2,5-Dimethylfuran

CAS Number: 625-86-5
MDL Number: MFCD00003250
Molecular Formula: C6H8O
Molecular Weight: 96.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): 16
Specific Gravity (20/20): 0.90

Formula: C₆H₈O
Boiling Pt: 93 °C
Density: 0.90 (20 °C)
Flash Pt: 16 °C
MDL Number: MFCD00003250
CAS Number: 625-86-5
UN: 3271
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/56/97/28975697.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR