Diisoamylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Diisopentylamine
D0613-25ML
TCD0613-25MLEA
408.84
USD
TCD0613-25ML
Diisoamylamine ≥97.0% (by GC, titration analysis)
Diisoamylamine
CAS Number: 544-00-3
MDL Number: MFCD00015053
Molecular Formula: C10H23N
Molecular Weight: 157.30
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 187
Flash Point (°C): 58
Specific Gravity (20/20): 0.77
Other Product Names / Synonyms: Diisopentylamine
Formula:
C₁₀H₂₃N MW: 157.30 g/mol Boiling Pt: 187 °C Density: 0.77 (20 °C) Flash Pt: 58 °C |
MDL Number:
MFCD00015053 CAS Number: 544-00-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...