Diethyl disulfide ≥99.0% (by GC)
Supplier: TCI America
D0486-25G
TCD0486-25GEA
51.77
USD
TCD0486-25G
Diethyl disulfide ≥99.0% (by GC)
Diethyl disulphide
CAS Number: 110-81-6
MDL Number: MFCD00009266
Molecular Formula: C4H10S2
Molecular Weight: 122.24
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 40
Specific Gravity (20/20): 0.99
Formula:
C₄H₁₀S₂ MW: 122.26 g/mol Boiling Pt: 154 °C Density: 0.99 (20 °C) Flash Pt: 40 °C |
MDL Number:
MFCD00009266 CAS Number: 110-81-6 UN: 1993 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...