1,3-Diethylbenzene ≥97.0% (by GC)

Supplier: TCI America

D0478-25ML D0478-100ML
TCD0478-25MLEA 91 USD
TCD0478-25ML TCD0478-100ML
1,3-Diethylbenzene ≥97.0% (by GC)
1,3-Diethylbenzene

CAS Number: 141-93-5
MDL Number: MFCD00009260
Molecular Formula: C10H14
Molecular Weight: 134.22
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 182
Melting point (°C): -84
Flash Point (°C): 56
Specific Gravity (20/20): 0.87

Formula: C₁₀H₁₄
MW: 134.22 g/mol
Boiling Pt: 182 °C
Melting Pt: –84 °C
Density: 0.87 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00009260
CAS Number: 141-93-5
UN: 2049
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/50/48/28975048.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR