2,5-Dichloro-p-xylene ≥97.0% (by GC)
Supplier: TCI America
D0431-25G
TCD0431-25GEA
91
USD
TCD0431-25G
2,5-Dichloro-p-xylene ≥97.0% (by GC)
2,5-Dichloro-p-xylene
CAS Number: 1124-05-6
MDL Number: MFCD00000610
Molecular Formula: C8H8Cl2
Molecular Weight: 175.05
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 108
Melting point (°C): 70
Formula:
C₈H₈Cl₂ MW: 175.06 g/mol Boiling Pt: 108 °C Melting Pt: 70 °C |
MDL Number:
MFCD00000610 CAS Number: 1124-05-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...