2,3-Dichloropropionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0407-5G D0407-25G
TCD0407-25GEA 170.29 USD
TCD0407-25G TCD0407-5G
2,3-Dichloropropionic acid ≥98.0% (by GC, titration analysis)
2,3-Dichloropropionic acid

CAS Number: 565-64-0
MDL Number: MFCD00021695
Molecular Formula: C3H4Cl2O2
Molecular Weight: 142.96
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 52

Formula: C₃H₄Cl₂O₂
MW: 142.96 g/mol
Boiling Pt: 210 °C
Melting Pt: 52 °C
MDL Number: MFCD00021695
CAS Number: 565-64-0
https://us.vwr.com/stibo/web/std.lang.all/49/31/28974931.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR