3,4-Dichlorophenol ≥98.0% (by GC)

Supplier: TCI America

D0395-10G
TCD0395-10GEA 163.42 USD
TCD0395-10G
3,4-Dichlorophenol ≥98.0% (by GC)
3,4-Dichlorophenol

CAS Number: 95-77-2
MDL Number: MFCD00002258
Molecular Formula: C6H4Cl2O
Molecular Weight: 163.00
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 118
Melting point (°C): 66

Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: 118 °C
Melting Pt: 66 °C
MDL Number: MFCD00002258
CAS Number: 95-77-2
UN: 2020
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/49/06/28974906.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR