2,6-Dichloro-4-nitrophenol ≥99.0% (by titrimetric analysis)

Supplier: TCI America

D0389-25G
TCD0389-25GEA 106.58 USD
TCD0389-25G
2,6-Dichloro-4-nitrophenol ≥99.0% (by titrimetric analysis)
2,6-Dichloro-4-nitrophenol

CAS Number: 618-80-4
MDL Number: MFCD00014715
Molecular Formula: C6H3Cl2NO3
Molecular Weight: 207.99
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 126

Formula: C₆H₃Cl₂NO₃
MW: 208 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00014715
CAS Number: 618-80-4
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/48/93/28974893.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR