trans-Dichloroethylene ≥98.0% (by GC) stabilized

Supplier: TCI America

Synonyms: trans-Dichloroethene(E)-1,2-Dichloroethenetrans-1,2-Dichloroethylene(E)-1,2-Dichloroethylene

D0368-25G
TCD0368-25GEA 81.77 USD
TCD0368-25G
trans-Dichloroethylene ≥98.0% (by GC) stabilized
trans-Dichloroethylene

(stabilized with MEHQ)
CAS Number: 156-60-5
MDL Number: MFCD00062942
Molecular Formula: C2H2Cl2
Molecular Weight: 96.94
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 48
Flash Point (°C): 2
Specific Gravity (20/20): 1.26


Other Product Names / Synonyms: trans-Acetylene Dichloride

Formula: C₂H₂Cl₂
MW: 96.94 g/mol
Boiling Pt: 48 °C
Density: 1.26 (20 °C)
Flash Pt: 2 °C
MDL Number: MFCD00062942
CAS Number: 156-60-5
UN: 1150
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/48/61/28974861.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR