TCIAD0366

D0366-250G
TCD0366-250GEA 65.08 USD
TCD0366-250G
TCIAD0366
(stabilized with MEHQ)
[To use this product charged in cylinder, a valve is required which is sold separately (Product Code:V0030)]
CAS Number: 75-35-4
MDL Number: MFCD00011653
Molecular Formula: C2H2Cl2
Molecular Weight: 96.94
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 31
Flash Point (°C): 10
Specific Gravity (20/20): 1.22

Product Categories: Synthetic Organic Chemistry, Gaseous Compounds, Materials Chemistry, Monomers
Other Product Names / Synonyms: Vinylidene Chloride
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR