1,3-Dichloro-2-butene ≥95.0% (by GC)
Supplier: TCI America
D0351-25G
TCD0351-25GEA
430.36
USD
TCD0351-25G
1,3-Dichloro-2-butene ≥95.0% (by GC)
1,3-Dichloro-2-butene
CAS Number: 926-57-8
MDL Number: MFCD00000861
Molecular Formula: C4H6Cl2
Molecular Weight: 124.99
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 27
Specific Gravity (20/20): 1.16
Formula:
C₄H₆Cl₂ MW: 125 g/mol Boiling Pt: 123 °C Density: 1.16 (20 °C) Flash Pt: 27 °C |
MDL Number:
MFCD00000861 CAS Number: 926-57-8 UN: 1992 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...