TCIAD0350

D0350-10G
TCD0350-10GEA 113.55 USD
TCD0350-10G
TCIAD0350
CAS Number: 7581-97-7
MDL Number: MFCD00000866
Molecular Formula: C4H8Cl2
Molecular Weight: 127.01
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 116
Flash Point (°C): 18
Specific Gravity (20/20): 1.12
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR