Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,2-Dichlorobutane ≥96.0% (by GC)

Supplier: TCI America

D0347-10G D0347-25G
TCD0347-10G TCD0347-25G
1,2-Dichlorobutane ≥96.0% (by GC)
CAS Number: 616-21-7
MDL Number: MFCD00013676
Molecular Formula: C4H8Cl2
Molecular Weight: 127.01
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 27
Specific Gravity (20/20): 1.12


Formula: ClH₂CCHClCH₂CH₃
MW: 127,01 g/mol
Boiling Pt: 124.1 °C (1013 hPa)
Density: 1.1116 g/cm³ (25 °C)
Flash Pt: 27 °C
MDL Number: MFCD00013676
CAS Number: 616-21-7
EINECS: 210-469-5
UN: 1992
ADR: 3,III
Order Now
Specification Test Results
Purity/Analysis Method >96.0% (GC)
Boiling point (°C) 124
Flash Point (°C) 27
Specific Gravity (20/20) 1.12


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty