3,4-Dichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D0346-25G
TCD0346-25GEA
38.97
USD
TCD0346-25G
3,4-Dichlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
3,4-Dichlorobenzoyl chloride
CAS Number: 3024-72-4
MDL Number: MFCD00000672
Molecular Formula: C7H3Cl3O
Molecular Weight: 209.45
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 242
Flash Point (°C): 142
Freezing point (°C): 30
Formula:
C₇H₃Cl₃O MW: 209.46 g/mol Boiling Pt: 242 °C Flash Pt: 142 °C |
MDL Number:
MFCD00000672 CAS Number: 3024-72-4 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...