Dichloramine B ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: N,N-DichlorobenzenesulphonamideBenzenesulfonedichloramideBenzenesulphonedichloramideN,N-Dichlorobenzenesulfonamide

D0317-25G D0317-5G
TCD0317-5GEA 82.18 USD
TCD0317-5G TCD0317-25G
Dichloramine B ≥95.0% (by titrimetric analysis)
Dichloramine B

CAS Number: 473-29-0
MDL Number: MFCD00025025
Molecular Formula: C6H5Cl2NO2S
Molecular Weight: 226.07
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 74


Other Product Names / Synonyms: Benzenesulfonedichloramide

Formula: C₆H₅Cl₂NO₂S
MW: 226.07 g/mol
Melting Pt: 74 °C
MDL Number: MFCD00025025
CAS Number: 473-29-0
https://us.vwr.com/stibo/web/std.lang.all/47/81/28974781.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR