2,5-Dichlorothiophene ≥97.0% (by GC)

Supplier: TCI America

D0250-25G
TCD0250-25GEA 107.02 USD
TCD0250-25G
2,5-Dichlorothiophene ≥97.0% (by GC)
2,5-Dichlorothiophene

CAS Number: 3172-52-9
MDL Number: MFCD00005423
Molecular Formula: C4H2Cl2S
Molecular Weight: 153.02
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Flash Point (°C): 59
Specific Gravity (20/20): 1.44
Storage Temperature: 0-10°C

Formula: C₄H₂Cl₂S
MW: 153.03 g/mol
Boiling Pt: 162 °C
Density: 1.44 (20 °C)
Flash Pt: 59 °C
MDL Number: MFCD00005423
CAS Number: 3172-52-9
UN: 2929
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/46/88/28974688.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR