α,α'-Dibromo-o-xylene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: o-Xylylene bromidealpha,alpha'-Dibromo-o-xyleneo-Xylylene dibromideo-Xylene dibromide1,2-Bis(bromomethyl)benzene

D0214-25G
TCD0214-25GEA 87.01 USD
TCD0214-25G
α,α'-Dibromo-o-xylene ≥98.0% (by GC)
α,α'-Dibromo-o-xylene

CAS Number: 91-13-4
MDL Number: MFCD00000175
Molecular Formula: C8H8Br2
Molecular Weight: 263.96
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 140
Melting point (°C): 93


Other Product Names / Synonyms: o-Xylylene Bromide, o-Xylylene Dibromide, 1,2-Bis(bromomethyl)benzene

Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 140 °C
Melting Pt: 93 °C
MDL Number: MFCD00000175
CAS Number: 91-13-4
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/46/35/28974635.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR