1,8-Diaminooctane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,8-Octanediamine Octamethylendiamine

D0107-25G
TCD0107-25GEA 30.62 USD
TCD0107-25G
1,8-Diaminooctane ≥98.0% (by GC, titration analysis)
1,8-Diaminooctane

CAS Number: 373-44-4
MDL Number: MFCD00008248
Molecular Formula: C8H20N2
Molecular Weight: 144.26
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 240
Melting point (°C): 50
Flash Point (°C): 114


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Octamethylenediamine, 1,8-Octanediamine

Formula: C₈H₂₀N₂
MW: 144.26 g/mol
Boiling Pt: 240 °C
Melting Pt: 50 °C
Flash Pt: 114 °C
MDL Number: MFCD00008248
CAS Number: 373-44-4
UN: 3259
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/44/77/28974477.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR