4H-Cyclopenta[2,1-b:3,4-b']dithiophene ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
4H-Cyclopenta[1,2-b:5,4-b']dithiophene
CAS Number: 389-58-2
MDL Number: MFCD20133900
Molecular Formula: C9H6S2
Molecular Weight: 178.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 73
Lambda max.: 305 nm (EtOH)Storage Temperature: 0-10°C
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors
Other Product Names / Synonyms: 4H-Cyclopenta[1,2-b:5,4-b']dithiophene
Formula:
C₉H₆S₂ MW: 178.27 g/mol Melting Pt: 73 °C |
MDL Number:
MFCD20133900 CAS Number: 389-58-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...