Chlormethiazole hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 5-(2-Chloroethyl)-4-methylthiazole HydrochlorideClomethiazole HydrochlorideSomnevrin Hydrochloride

C3020-200MG C3020-1G
TCC3020-200MGEA 194.88 USD
TCC3020-200MG TCC3020-1G
Chlormethiazole hydrochloride ≥98.0% (by HPLC, titration analysis)
Chlormethiazole hydrochloride

CAS Number: 6001-74-7
MDL Number: MFCD00673948
Molecular Formula: C6H8ClNS
Molecular Weight: 198.11
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 136


Product Categories: Bioscience, Neurotransmitter Agent Ingredients
Other Product Names / Synonyms: 5-(2-Chloroethyl)-4-methylthiazole Hydrochloride, Clomethiazole Hydrochloride, Somnevrin Hydrochloride

Formula: C₆H₈ClNS
MW: 198.11 g/mol
Melting Pt: 136 °C
MDL Number: MFCD00673948
CAS Number: 6001-74-7
https://us.vwr.com/stibo/web/std.lang.all/70/79/28967079.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR