Cy-cBRIDP® ≥95.0% (by HPLC)
Synonyms:
Dicyclohexyl(1-methyl-2,2-diphenylcyclopropyl)phosphine
CAS Number: 1023330-38-2
MDL Number: MFCD10565626
Molecular Formula: C28H37P
Molecular Weight: 404.58
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 122
Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: Dicyclohexyl(1-methyl-2,2-diphenylcyclopropyl)phosphine
Formula:
C₂₈H₃₇P MW: 404.58 g/mol Melting Pt: 122 °C |
MDL Number:
MFCD10565626 CAS Number: 1023330-38-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...