Cyclopentyl isocyanide ≥98.0% (by GC)

Supplier: TCI America

C2959-1G
TCC2959-1GEA 225.33 USD
TCC2959-1G
Cyclopentyl isocyanide ≥98.0% (by GC)
Cyclopentyl isocyanide

CAS Number: 68498-54-4
MDL Number: MFCD02664624
Molecular Formula: C6H9N
Molecular Weight: 95.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 28
Specific Gravity (20/20): 0.89
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Isonitriles (Multi-component Reaction)
Other Product Names / Synonyms: Isocyanocyclopentane

Formula: C₆H₉N
Boiling Pt: 28 °C
Density: 0.89 (20 °C)
MDL Number: MFCD02664624
CAS Number: 68498-54-4
UN: 2810
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/70/24/28967024.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR