1-(Chloroacetyl)pyrrolidine ≥97.0%

Supplier: TCI America

Synonyms: 2-Chloro-1-(pyrrolidin-1-yl)ethanone

C2948-1G C2948-5G
TCC2948-5GEA 307 USD
TCC2948-5G TCC2948-1G
1-(Chloroacetyl)pyrrolidine ≥97.0%
1-(Chloroacetyl)pyrrolidine

CAS Number: 20266-00-6
MDL Number: MFCD01217237
Molecular Formula: C6H10ClNO
Molecular Weight: 147.60
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Boiling point (°C): 105
Melting point (°C): 46

Formula: C₆H₁₀ClNO
MW: 147.60 g/mol
Boiling Pt: 105 °C
Melting Pt: 46 °C
MDL Number: MFCD01217237
CAS Number: 20266-00-6
https://us.vwr.com/stibo/web/std.lang.all/70/15/28967015.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR