2-Chloro-1,1,2-trifluoroethyl ethyl ether ≥98.0% (by GC)

Supplier: TCI America

C2862-5G
TCC2862-5GEA 74 USD
TCC2862-5G
2-Chloro-1,1,2-trifluoroethyl ethyl ether ≥98.0% (by GC)
2-Chloro-1,1,2-trifluoroethyl ethyl ether

CAS Number: 310-71-4
MDL Number: MFCD00018849
Molecular Formula: C4H6ClF3O
Molecular Weight: 162.54
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Specific Gravity (20/20): 1.23

Formula: C₄H₆ClF₃O
MW: 162.54 g/mol
Boiling Pt: 88 °C
Density: 1.23 (20 °C)
MDL Number: MFCD00018849
CAS Number: 310-71-4
https://us.vwr.com/stibo/web/std.lang.all/69/53/28966953.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR