2-Chloro-3-(chloromethyl)quinoline ≥98.0% (by GC)

Supplier: TCI America

C2861-5G C2861-1G
TCC2861-1GEA 115 USD
TCC2861-1G TCC2861-5G
2-Chloro-3-(chloromethyl)quinoline ≥98.0% (by GC)
2-Chloro-3-(chloromethyl)quinoline

CAS Number: 90097-52-2
MDL Number: MFCD05865134
Molecular Formula: C10H7Cl2N
Molecular Weight: 212.07
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 117

https://us.vwr.com/stibo/web/std.lang.all/69/52/28966952.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR