Chitosan oligosaccharides

Supplier: TCI America

C2849-25G C2849-100G
TCC2849-100GEA 368.34 USD
TCC2849-100G TCC2849-25G
Chitosan oligosaccharides
Chitosan oligosaccharides

Color: Slightly Pale Yellow
Form: Crystal


Product Categories: Bioscience, Oligosaccharides

MDL Number: MFCD00161512
CAS Number: 9012-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR