(Carbobenzoxy)glycylglycine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Z-Gly-Gly-OH

C2833-5G
TCC2833-5GEA 234.27 USD
TCC2833-5G
(Carbobenzoxy)glycylglycine ≥98.0% (by HPLC, titration analysis)
(Carbobenzoxy)glycylglycine

CAS Number: 2566-19-0
MDL Number: MFCD00037783
Molecular Formula: C12H14N2O5
Molecular Weight: 266.25
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 180


Product Categories: Bioscience, Oligopeptides
Other Product Names / Synonyms: Z-Gly-Gly-OH

Formula: C₁₂H₁₄N₂O₅
MW: 266.25 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00037783
CAS Number: 2566-19-0
https://us.vwr.com/stibo/web/std.lang.all/69/34/28966934.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR