Cyclic pifithrin-ɑ-hydrobromide ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-(p-Tolyl)-5,6,7,8-tetrahydroimidazo[2,1-b]benzothiazole Hydrobromide

C2826-20MG C2826-100MG
TCC2826-100MGEA 1006 USD
TCC2826-100MG TCC2826-20MG
Cyclic pifithrin-ɑ-hydrobromide ≥98.0% (by HPLC)
Cyclic pifithrin-alpha-hydrobromide

CAS Number: 511296-88-1
MDL Number: MFCD02683960
Molecular Formula: C16H16N2S
Molecular Weight: 349.29
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Storage Temperature: 0°C


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Regenerative Medicine Research Reagents
Other Product Names / Synonyms: 2-(p-Tolyl)-5,6,7,8-tetrahydroimidazo[2,1-b]benzothiazole Hydrobromide

Formula: C₁₆H₁₆N₂S
MW: 349.29 g/mol
MDL Number: MFCD02683960
CAS Number: 511296-88-1
https://us.vwr.com/stibo/web/std.lang.all/69/28/28966928.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR