2-Cyano-N,N-diethylacetamide ≥98.0% (by GC)

Supplier: TCI America

C2822-25G C2822-5G
TCC2822-5GEA 71.03 USD
TCC2822-5G TCC2822-25G
2-Cyano-N,N-diethylacetamide ≥98.0% (by GC)
2-Cyano-N,N-diethylacetamide

CAS Number: 26391-06-0
MDL Number: MFCD01332383
Molecular Formula: C7H12N2O
Molecular Weight: 140.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 114
Flash Point (°C): 110
Specific Gravity (20/20): 1.02
Storage Temperature: 0-10°C

Formula: C₇H₁₂N₂O
MW: 140.19 g/mol
Boiling Pt: 114 °C
Density: 1.02 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01332383
CAS Number: 26391-06-0
https://us.vwr.com/stibo/web/std.lang.all/69/25/28966925.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR