Catecholbis(trifluoromethanesulfonate) ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Catechol DitriflateCatechol Bis(trifluoromethanesulphonate)Catechol Bis(trifluoromethanesulfonate)

C2809-5G C2809-1G
TCC2809-1GEA 167 USD
TCC2809-1G TCC2809-5G
Catecholbis(trifluoromethanesulfonate) ≥98.0% (by GC)
Catecholbis(trifluoromethanesulfonate)

CAS Number: 17763-91-6
MDL Number: MFCD00274274
Molecular Formula: C8H4F6O6S2
Molecular Weight: 374.22
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 95
Melting point (°C): 37
Flash Point (°C): 113


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives
Other Product Names / Synonyms: Catechol Ditriflate

Formula: C₈H₄F₆O₆S₂
MW: 374.22 g/mol
Boiling Pt: 95 °C
Melting Pt: 37 °C
Flash Pt: 113 °C
MDL Number: MFCD00274274
CAS Number: 17763-91-6
https://us.vwr.com/stibo/web/std.lang.all/69/15/28966915.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR