5-[4-[3-Chloro-4-(3-fluorobenzyloxy)anilino]-6-quinazolinyl]furan-2-carboxaldehyde ≥97.0% (by HPLC, titration analysis)
Synonyms:
5-(4-((3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)amino)quinazolin-6-yl)furan-2-carbaldehyde
C2786-1G
C2786-5G
TCC2786-5GEA
192.61
USD
TCC2786-5G
TCC2786-1G
5-[4-[3-Chloro-4-(3-fluorobenzyloxy)anilino]-6-quinazolinyl]furan-2-carboxaldehyde ≥97.0% (by HPLC, titration analysis)
5-[4-[3-Chloro-4-(3-fluorobenzyloxy)anilino]-6-quinazolinyl]furan-2-carboxaldehyde
CAS Number: 231278-84-5
MDL Number: MFCD10565688
Molecular Formula: C26H17ClFN3O3
Molecular Weight: 473.89
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 229
Formula:
C₂₆H₁₇ClFN₃O₃ MW: 473.89 g/mol Melting Pt: 229 °C |
MDL Number:
MFCD10565688 CAS Number: 231278-84-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...