Cephalothin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Cephalotin7-(2-Thienylacetamido)cephalosporanic Acid

C2769-5G C2769-25G
TCC2769-5GEA 84.12 USD
TCC2769-5G TCC2769-25G
Cephalothin ≥98.0% (by HPLC, titration analysis)
Cephalothin

CAS Number: 153-61-7
MDL Number: MFCD00242614
Molecular Formula: C16H16N2O6S2
Molecular Weight: 396.43
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 154
Lambda max.: 263 nm (H2O)


Product Categories: Bioscience, Antibiotic Ingredients
Other Product Names / Synonyms: Cephalotin, 7-(2-Thienylacetamido)cephalosporanic Acid

Formula: C₁₆H₁₆N₂O₆S₂
MW: 396.43 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00242614
CAS Number: 153-61-7
https://us.vwr.com/stibo/web/std.lang.all/68/85/28966885.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR