Cyclohexanecarboxylic acid anhydride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
Cyclohexanecarboxylic Anhydride
C2735-5G
TCC2735-5GEA
164.43
USD
TCC2735-5G
Cyclohexanecarboxylic acid anhydride ≥98.0% (by GC, titration analysis)
Cyclohexanecarboxylic acid anhydride
CAS Number: 22651-87-2
MDL Number: MFCD12404944
Molecular Formula: C14H22O3
Molecular Weight: 238.33
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 156
Specific Gravity (20/20): 1.05
Formula:
C₁₄H₂₂O₃ MW: 238.3227 g/mol Boiling Pt: 156 °C Density: 1.05 (20 °C) |
MDL Number:
MFCD12404944 CAS Number: 22651-87-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...