4-Chloropyridine-2-carbonitrile ≥98.0% (by GC)

Supplier: TCI America

C2732-1G C2732-5G
TCC2732-1GEA 41.55 USD
TCC2732-1G TCC2732-5G
4-Chloropyridine-2-carbonitrile ≥98.0% (by GC)
4-Chloropyridine-2-carbonitrile

CAS Number: 19235-89-3
MDL Number: MFCD06009833
Molecular Formula: C6H3ClN2
Molecular Weight: 138.55
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: 4-Chloro-2-pyridinecarbonitrile, 4-Chloropicolinonitrile

Formula: C₆H₃ClN₂
MW: 138.5500 g/mol
Melting Pt: 84 °C
MDL Number: MFCD06009833
CAS Number: 19235-89-3
https://us.vwr.com/stibo/web/std.lang.all/68/55/28966855.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR