Chloro(dimethylsulfide)gold(I) ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (Dimethylsulfide)gold(I) Chloride(Dimethylsulphide)gold(I) Chloride

C2719-200MG C2719-1G
TCC2719-200MGEA 104.51 USD
TCC2719-200MG TCC2719-1G
Chloro(dimethylsulfide)gold(I) ≥95.0% (by titrimetric analysis)
Chloro(dimethylsulfide)gold(I)

CAS Number: 29892-37-3
MDL Number: MFCD00192090
Molecular Formula: C2H6AuClS
Molecular Weight: 294.55
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 160
Storage Temperature: 0-10°C


Other Product Names / Synonyms: (Dimethylsulfide)gold(I) Chloride

Formula: C₂H₆AuClS
MW: 294.55 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00192090
CAS Number: 29892-37-3
https://us.vwr.com/stibo/web/std.lang.all/68/44/28966844.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR