1-Cyclohexenylboronic acid pinacol ester ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane1-Cyclohexene-1-boronic acid pinacol ester

C2707-1G
TCC2707-1GEA 137.03 USD
TCC2707-1G
1-Cyclohexenylboronic acid pinacol ester ≥95.0% (by GC)
1-Cyclohexenylboronic acid pinacol ester

CAS Number: 141091-37-4
MDL Number: MFCD05663845
Molecular Formula: C12H21BO2
Molecular Weight: 208.11
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 103
Specific Gravity (20/20): 0.96
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Boronic Acids & Boronic Acid Derivatives
Other Product Names / Synonyms: 1-Cyclohexeneboronic Acid Pinacol Ester, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclohexene

Formula: C₁₂H₂₁BO₂
MW: 208.1049 g/mol
Density: 0.96 (20 °C)
Flash Pt: 103 °C
MDL Number: MFCD05663845
CAS Number: 141091-37-4
https://us.vwr.com/stibo/web/std.lang.all/68/33/28966833.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR