4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Chloro-3-(trifluoromethyl)benzene-1-sulphonyl chloride

C2705-5G C2705-1G
TCC2705-5GEA 503 USD
TCC2705-5G TCC2705-1G
4-Chloro-3-(trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
4-Chloro-3-(trifluoromethyl)benzenesulphonyl chloride

CAS Number: 32333-53-2
MDL Number: MFCD00024882
Molecular Formula: C7H3Cl2F3O2S
Molecular Weight: 279.05
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 131
Freezing point (°C): 28

Formula: C₇H₃Cl₂F₃O₂S
MW: 279.07 g/mol
Boiling Pt: 131 °C
MDL Number: MFCD00024882
CAS Number: 32333-53-2
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/68/31/28966831.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR