Chlorodimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Dimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silyl Chloride1-[3-(Chlorodimethylsilyl)propyl]-2,3,4,5,6-pentafluorobenzene

C2700-5G
TCC2700-5GEA 223.24 USD
TCC2700-5G
Chlorodimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane ≥95.0% (by GC)
Chlorodimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silane

CAS Number: 157499-19-9
MDL Number: MFCD00054911
Molecular Formula: C11H12ClF5Si
Molecular Weight: 302.74
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 73
Specific Gravity (20/20): 1.28


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Dimethyl[3-(2,3,4,5,6-pentafluorophenyl)propyl]silyl Chloride, 1-[3-(Chlorodimethylsilyl)propyl]-2,3,4,5,6-pentafluorobenzene

Formula: C₁₁H₁₂ClF₅Si
MW: 302.74 g/mol
Boiling Pt: 73 °C
Density: 1.28 (20 °C)
MDL Number: MFCD00054911
CAS Number: 157499-19-9
https://us.vwr.com/stibo/web/std.lang.all/68/25/28966825.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR