N-(Cyclopentyloxycarbonyloxy)succinimide ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Cyclopentyl N-Succinimidyl Carbonate

C2695-25G
TCC2695-25GEA 94 USD
TCC2695-25G
N-(Cyclopentyloxycarbonyloxy)succinimide ≥98.0% (by GC)
N-(Cyclopentyloxycarbonyloxy)succinimide

CAS Number: 128595-07-3
MDL Number: MFCD04038113
Molecular Formula: C10H13NO5
Molecular Weight: 227.22
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 80
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Cyclopentyl N-Succinimidyl Carbonate

Formula: C₁₀H₁₃NO₅
MW: 227.22 g/mol
Melting Pt: 80 °C
MDL Number: MFCD04038113
CAS Number: 128595-07-3
https://us.vwr.com/stibo/web/std.lang.all/68/20/28966820.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR