3-Cyano-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
3-Cyano-4-fluorobenzeneboronic acid
C2676-1G
TCC2676-1GEA
50.75
USD
TCC2676-1G
3-Cyano-4-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Cyano-4-fluorophenylboronic acid
CAS Number: 214210-21-6
MDL Number: MFCD03095130
Molecular Formula: C7H5BFNO2
Molecular Weight: 164.93
Form: Crystal
Color: White
Other Product Names / Synonyms: 3-Cyano-4-fluorobenzeneboronic Acid
Formula:
C₇H₅BFNO₂ MW: 164.93 g/mol |
MDL Number:
MFCD03095130 CAS Number: 214210-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...