2-Chlorotropone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Chlorocyclohepta-2,4,6-trienone2-Chloro-2,4,6-cycloheptatrien-1-one

C2674-1G
TCC2674-1GEA 202.94 USD
TCC2674-1G
2-Chlorotropone ≥98.0% (by GC)
2-Chlorotropone

CAS Number: 3839-48-3
MDL Number: MFCD00182497
Molecular Formula: C7H5ClO
Molecular Weight: 140.57
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 97
Melting point (°C): 68


Other Product Names / Synonyms: 2-Chloro-2,4,6-cycloheptatrien-1-one

Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 97 °C
Melting Pt: 68 °C
MDL Number: MFCD00182497
CAS Number: 3839-48-3
https://us.vwr.com/stibo/web/std.lang.all/97/25/28939725.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR